Assay Details
Gene Symbol: Parvg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130583.1 | NP_001124055.1 | 7 - 8 | 694 | 101 | |
XM_006241411.1 | XP_006241473.1 | 7 - 8 | 651 | 101 | ||
XM_006241412.2 | XP_006241474.1 | 7 - 8 | 904 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
