Assay Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244855.1 | NP_001231784.1 | 3 - 4 | 455 | 67 | |
XM_006242100.2 | XP_006242162.1 | 3 - 4 | 1895 | 67 | ||
XM_006242101.2 | XP_006242163.1 | 3 - 4 | 744 | 67 | ||
GenBank mRNA | BC082056.1 | - | 3 - 4 | 433 | 7132303 | 67 |
CV126932.1 | - | 3 - 4 | 455 | 67 | ||
FQ217036.1 | - | 3 - 4 | 439 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
