Assay Details
Gene Symbol: Rnf19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130560.1 | NP_001124032.1 | 9 - 10 | 2070 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P150814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413709 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127822 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150814 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P150814 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P150814 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P16696 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P401284 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10903322 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903323 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903324 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903317 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903316 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903319 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903318 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903321 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903320 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10903315 | - | - |
Rat Genome 230 2.0 Array | 1378524_at | - | - |
Rat Genome 230 2.0 Array | 1388937_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388937_at | - | - |
Rat Genome 230 2.0 Array | 1378524_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378524_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378524_at | - | - |
Affymetrix HT Rat Focus Array | 1388937_at | - | - |
Affymetrix HT Rat Focus Array | 1388937_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700023094.rn.1 | TC0700002983.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700023095.rn.1 | TC0700002983.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700023096.rn.1 | TC0700002983.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700023093.rn.1 | TC0700002983.rn.1 | RTA TC |
Rat Expression Set 230 | 1388937_at | - | - |
Rat Expression Set 230 | 1388937_PM_at | - | - |
Rat Expression Set 230 | 1378524_PM_at | - | - |
Rat Expression Set 230 | 1378524_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10903315 | - | - |
Rat Genome U34 Set | rc_AI044672_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21826121 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367545 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649779 | - | - |
