Assay Details
Gene Symbol: Ift27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130495.1 | NP_001123967.1 | 5 - 6 | 654 | 63 | |
XM_006241931.2 | XP_006241993.1 | 5 - 6 | 666 | 63 | ||
GenBank mRNA | BC168150.1 | - | 664 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
