Assay Details
Gene Symbol: Adsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130503.1 | NP_001123975.1 | 12 - 13 | 1407 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
purine nucleotide metabolic process
AMP biosynthetic process
'de novo' IMP biosynthetic process
response to nutrient
metabolic process
aerobic respiration
ribonucleoside monophosphate biosynthetic process
response to muscle activity
response to starvation
'de novo' AMP biosynthetic process
protein tetramerization
N6-(1,2-dicarboxyethyl)AMP AMP-lyase (fumarate-forming) activity
(S)-2-(5-amino-1-(5-phospho-D-ribosyl)imidazole-4-carboxamido)succinate AMP-lyase (fumarate-forming) activity
response to hypoxia
purine nucleotide metabolic process
AMP biosynthetic process
'de novo' IMP biosynthetic process
response to nutrient
metabolic process
aerobic respiration
ribonucleoside monophosphate biosynthetic process
response to muscle activity
response to starvation
'de novo' AMP biosynthetic process
protein tetramerization
N6-(1,2-dicarboxyethyl)AMP AMP-lyase (fumarate-forming) activity
(S)-2-(5-amino-1-(5-phospho-D-ribosyl)imidazole-4-carboxamido)succinate AMP-lyase (fumarate-forming) activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921181 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458270 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3213 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11430 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11430 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11430 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11430 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10897862 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897855 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897858 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897856 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897857 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897864 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897866 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897865 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897867 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897863 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897861 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897860 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10897854 | - | - |
Rat Genome 230 2.0 Array | 1376091_PM_at | - | - |
Rat Genome 230 2.0 Array | 1376091_at | - | - |
Affymetrix HT Rat Focus Array | 1376091_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1376091_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0700011235.rn.1 | TC0700001486.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0700011236.rn.1 | TC0700001486.rn.1 | RTA TC |
Rat Expression Set 230 | 1376091_PM_at | - | - |
Rat Expression Set 230 | 1376091_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10897854 | - | - |
Rat Genome U34 Set | rc_AI172062_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21802445 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369783 | - | - |
