Assay Details
Gene Symbol: Aldob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012496.2 | NP_036628.2 | 6 - 7 | 683 | 84 | |
GenBank mRNA | BC081697.1 | - | 6 - 7 | 683 | 84 | |
FQ209505.1 | - | 6 - 7 | 703 | 84 | ||
FQ209583.1 | - | 6 - 7 | 703 | 84 | ||
FQ209639.1 | - | 6 - 7 | 703 | 84 | ||
FQ209643.1 | - | 6 - 7 | 700 | 84 | ||
FQ209782.1 | - | 6 - 7 | 703 | 84 | ||
FQ210002.1 | - | 6 - 7 | 703 | 84 | ||
FQ210849.1 | - | 6 - 7 | 703 | 84 | ||
FQ211156.1 | - | 6 - 7 | 703 | 84 | ||
FQ211159.1 | - | 6 - 7 | 703 | 84 | ||
FQ211293.1 | - | 4 - 5 | 381 | 84 | ||
FQ218139.1 | - | 6 - 7 | 703 | 84 | ||
FQ218348.1 | - | 6 - 7 | 709 | 84 | ||
FQ218522.1 | - | 6 - 7 | 703 | 84 | ||
FQ218551.1 | - | 6 - 7 | 703 | 84 | ||
FQ218592.1 | - | 2 - 3 | 201 | 84 | ||
FQ218665.1 | - | 6 - 7 | 703 | 84 | ||
FQ218885.1 | - | 6 - 7 | 702 | 84 | ||
FQ218911.1 | - | 6 - 7 | 703 | 84 | ||
FQ219077.1 | - | 4 - 5 | 387 | 84 | ||
FQ219096.1 | - | 6 - 7 | 703 | 84 | ||
FQ219116.1 | - | 6 - 7 | 703 | 84 | ||
FQ219236.1 | - | 6 - 7 | 702 | 84 | ||
FQ219261.1 | - | 6 - 7 | 703 | 84 | ||
FQ219555.1 | - | 6 - 7 | 703 | 84 | ||
FQ219574.1 | - | 6 - 7 | 703 | 84 | ||
FQ219585.1 | - | 6 - 7 | 703 | 84 | ||
FQ219640.1 | - | 6 - 7 | 701 | 84 | ||
FQ219646.1 | - | 6 - 7 | 703 | 84 | ||
FQ219703.1 | - | 6 - 7 | 702 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
