Assay Details
Gene Symbol: Naaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010967.2 | NP_001010967.1 | 6 - 7 | 871 | 64 | |
GenBank mRNA | AB162193.1 | - | 6 - 7 | 869 | 64 | |
BC105771.1 | - | 6 - 7 | 871 | 7930474 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
