Assay Details
Gene Symbol: Fkbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012174.1 | NP_001012174.1 | 4 - 5 | 503 | 90 | |
XM_006256222.2 | XP_006256284.1 | 4 - 5 | 1252 | 90 | ||
XM_006256224.2 | XP_006256286.1 | 5 - 6 | 1319 | 90 | ||
GenBank mRNA | BC085868.1 | - | 4 - 5 | 503 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
