Assay Details
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256148.1 | XP_006256210.1 | 2 - 3 | 253 | 117 | |
XM_006256149.1 | XP_006256211.1 | 2 - 3 | 244 | 117 | ||
XM_228042.6 | XP_228042.4 | 1 - 2 | 462 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
