Assay Details
Gene Symbol: Slc25a16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100860.1 | NP_001094330.1 | 3 - 4 | 427 | 127 | |
GenBank mRNA | BC166494.1 | - | 3 - 4 | 427 | 127 | |
M32973.1 | - | 3 - 4 | 431 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
