Assay Details
Gene Symbol: Rnf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025727.1 | NP_001020898.1 | 1 - 2 | 198 | 72 | |
XM_006256225.2 | XP_006256287.1 | 1 - 2 | 275 | 72 | ||
XM_006256226.2 | XP_006256288.1 | 1 - 2 | 278 | 72 | ||
GenBank mRNA | BC099079.1 | - | 1 - 2 | 198 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
