Assay Details
Gene Symbol: Dld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199385.2 | NP_955417.1 | 4 - 5 | 341 | 97 | |
XM_008764590.1 | XP_008762812.1 | 4 - 5 | 358 | 97 | ||
GenBank mRNA | BC062069.1 | - | 4 - 5 | 345 | 97 | |
FM100976.1 | - | 4 - 5 | 341 | 97 | ||
FQ215299.1 | - | 4 - 5 | 348 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
