Assay Details
Gene Symbol: Acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135562.1 | NP_001129034.1 | 3 - 4 | 300 | 82 | |
XM_008764572.1 | XP_008762794.1 | 3 - 4 | 300 | 82 | ||
GenBank mRNA | BC158763.1 | - | 3 - 4 | 300 | 7109165 | 82 |
BC161990.1 | - | 3 - 4 | 246 | 9032684 | 82 | |
FQ215147.1 | - | 3 - 4 | 238 | 82 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
