Assay Details
Gene Symbol: Cenpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750123.2 | XP_003750171.1 | 4 - 5 | 625 | 69 | |
XM_006239855.1 | XP_006239917.1 | 4 - 5 | 625 | 69 | ||
XM_008764544.1 | XP_008762766.1 | 4 - 5 | 691 | 69 | ||
XM_008764545.1 | XP_008762767.1 | 4 - 5 | 691 | 69 | ||
XM_008764546.1 | XP_008762768.1 | 4 - 5 | 691 | 69 | ||
GenBank mRNA | FQ233391.1 | - | 2 - 3 | 588 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
