Assay Details
Gene Symbol: Mfsd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729615.3 | XP_002729661.1 | 10 - 11 | 1372 | 68 | |
XM_006239869.2 | XP_006239931.1 | 10 - 11 | 1399 | 68 | ||
XM_006239870.2 | XP_006239932.1 | 9 - 10 | 1273 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
