Assay Details
Gene Symbol: Pqlc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034952.1 | NP_001030124.1 | 2 - 3 | 393 | 78 | |
XM_008764587.1 | XP_008762809.1 | 2 - 3 | 716 | 78 | ||
GenBank mRNA | CB556892.1 | - | 2 - 3 | 547 | 78 | |
CK477351.1 | - | 2 - 3 | 216 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
