Assay Details
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106740.1 | NP_001100210.1 | 5 - 6 | 1082 | 78 | |
XM_006240222.2 | XP_006240284.1 | 5 - 6 | 1132 | 78 | ||
XM_006240223.2 | XP_006240285.1 | 5 - 6 | 1130 | 78 | ||
XM_006240224.2 | XP_006240286.1 | 4 - 5 | 870 | 78 | ||
XM_006240225.2 | XP_006240287.1 | 4 - 5 | 779 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
