Assay Details
Gene Symbol: Gucy1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012769.2 | NP_036901.2 | 1 - 2 | 93 | 87 | |
GenBank mRNA | BC081840.1 | - | 1 - 2 | 93 | 7104600 | 87 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC081840.1,NM_012769.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
