Assay Details
Gene Symbol: Slc25a44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108947.1 | NP_001102417.1 | 2 - 3 | 1031 | 91 | |
XM_006232719.2 | XP_006232781.1 | 2 - 3 | 866 | 91 | ||
XM_006232720.2 | XP_006232782.1 | 2 - 3 | 1022 | 91 | ||
XM_006232721.1 | XP_006232783.1 | 2 - 3 | 891 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
