Assay Details
Gene Symbol: Npm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203340.1 | NP_976085.1 | 7 - 8 | 577 | 71 | |
XM_006252261.2 | XP_006252323.1 | 8 - 9 | 1387 | 71 | ||
XM_008770824.1 | XP_008769046.1 | 3 - 4 | 1213 | 71 | ||
XM_008770825.1 | XP_008769047.1 | 4 - 5 | 1266 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
