Assay Details
Gene Symbol: Edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191899.1 | NP_001178828.1 | 8 - 9 | 804 | 67 | |
XM_006256381.2 | XP_006256443.1 | 9 - 10 | 1097 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
