Assay Details
Gene Symbol: Edar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191899.1 | NP_001178828.1 | 10 - 11 | 1021 | 120 | |
XM_006256381.2 | XP_006256443.1 | 11 - 12 | 1314 | 120 | ||
GenBank mRNA | FQ211923.1 | - | 11 - 12 | 1245 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
