Assay Details
Gene Symbol: LOC679863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762423.1 | XP_008760645.1 | 3 - 4 | 303 | 75 |
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013923.1 | NP_001013945.1 | 3 - 4 | 258 | 75 | |
GenBank mRNA | BC078946.1 | - | 3 - 4 | 258 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P002870 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P281435 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1058916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73913 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1058916 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002870 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P281435 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1058916 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P281435 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P002870 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P281435 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1058916 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P002870 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P281435 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1058916 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P268837 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10765 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54265 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31228 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83351 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5221 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66152 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 18034 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 47822 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37271 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10851215 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851221 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851225 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851220 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851229 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851216 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851212 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851222 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851223 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851226 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10851210 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10851219 | - | - |
Rat Genome 230 2.0 Array | 1370903_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1370903_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300035383.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035387.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035395.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035378.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035393.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035404.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035412.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035373.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035394.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035402.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035376.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035377.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035388.rn.1 | TC0300004595.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300035407.rn.1 | TC0300004595.rn.1 | RTA TC |
Rat Expression Set 230 | 1370903_PM_a_at | - | - |
Rat Expression Set 230 | 1370903_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851210 | - | - |
Rat Genome U34 Set | M59814_at | - | - |
Rat Genome U34 Set | rc_AI230982_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21050353 | - | - |
ABI Rat Genome Survey Microarray | 21606569 | - | - |
