Assay Details
Gene Symbol: Blmh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034163.1 | NP_001029335.1 | 3 - 4 | 496 | 149 | |
GenBank mRNA | BC128701.1 | - | 3 - 4 | 381 | 7379958 | 149 |
CK363809.1 | - | 3 - 4 | 423 | 149 | ||
D87336.1 | - | 3 - 4 | 399 | 149 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
