Assay Details
Gene Symbol: Foxred2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191787.1 | NP_001178716.1 | 1 - 2 | 496 | 81 | |
XM_006241932.2 | XP_006241994.1 | 2 - 3 | 1542 | 81 | ||
XM_008765712.1 | XP_008763934.1 | 2 - 3 | 1453 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
