Assay Details
Gene Symbol: Gpatch8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001062937.5 | XP_001062937.3 | 1 - 2 | 269 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
