Assay Details
Gene Symbol: Gpatch8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001062937.5 | XP_001062937.3 | 2 - 3 | 340 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
