Assay Details
Gene Symbol: Gpatch8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001062937.5 | XP_001062937.3 | 3 - 4 | 414 | 78 | |
XM_008768226.1 | XP_008766448.1 | 1 - 2 | 1390 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
