Assay Details
Gene Symbol: Lancl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053723.1 | NP_446175.1 | 4 - 5 | 685 | 103 | |
GenBank mRNA | AJ131111.1 | - | 4 - 5 | 685 | 103 | |
BC085811.1 | - | 4 - 5 | 663 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
