Assay Details
Gene Symbol: Lancl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053723.1 | NP_446175.1 | 7 - 8 | 1200 | 71 | |
GenBank mRNA | AJ131111.1 | - | 7 - 8 | 1200 | 71 | |
BC085811.1 | - | 7 - 8 | 1178 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
