Assay Details
Gene Symbol: Akap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133515.2 | NP_598199.2 | 1 - 2 | 285 | 119 | |
GenBank mRNA | U67136.1 | - | 1 - 2 | 119 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_133515.2,U67136.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
