Assay Details
Gene Symbol: Slc1a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198763.1 | NP_942058.1 | 1 - 2 | 528 | 83 | |
GenBank mRNA | AB103401.1 | - | 1 - 2 | 528 | 83 | |
FQ210448.1 | - | 756 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
