Assay Details
Gene Symbol: Slc1a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198763.1 | NP_942058.1 | 4 - 5 | 803 | 72 | |
GenBank mRNA | AB103401.1 | - | 4 - 5 | 803 | 72 | |
FQ210448.1 | - | 1031 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
