Assay Details
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199119.1 | NP_954550.1 | 7 - 8 | 2215 | 83 | |
XM_006240483.1 | XP_006240545.1 | 7 - 8 | 2418 | 83 | ||
XM_006240484.2 | XP_006240546.1 | 7 - 8 | 2650 | 83 | ||
GenBank mRNA | AY341876.1 | - | 7 - 8 | 2215 | 83 | |
BC097262.1 | - | 7 - 8 | 2202 | 83 | ||
FQ212095.1 | - | 5 - 6 | 961 | 83 | ||
FQ218902.1 | - | 3 - 4 | 332 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
