Assay Details
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231757.2 | XP_006231819.1 | 2 | 1122 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
