Assay Details
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009539.1 | NP_001009539.1 | 8 | 1846 | 120 | |
XM_006250253.2 | XP_006250315.1 | 8 | 1877 | 120 | ||
XM_006250254.2 | XP_006250316.1 | 8 | 1997 | 120 | ||
GenBank mRNA | BC061812.1 | - | 8 | 1846 | 120 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
