Assay Details
Gene Symbol: Tomm40l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250247.2 | XP_006250309.1 | 10 | 99 |
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270838.1 | NP_001257767.1 | 9 | 1307 | 99 | |
NM_001270839.1 | NP_001257768.1 | 9 | 1285 | 99 | ||
NM_001270840.1 | NP_001257769.1 | 9 | 1304 | 99 | ||
NM_022941.4 | NP_075230.1 | 9 | 1289 | 99 | ||
XM_006250261.2 | XP_006250323.1 | 9 | 1395 | 99 | ||
XM_008769759.1 | XP_008767981.1 | 8 | 2252 | 99 | ||
GenBank mRNA | AB104736.1 | - | 9 | 1342 | 99 | |
AB105072.1 | - | 8 | 1433 | 99 | ||
BC089763.1 | - | 9 | 1304 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
