Assay Details
Gene Symbol: Tbata | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772957.1 | XP_008771179.1 | 9 | 2442 | 139 | |
XM_008772958.1 | XP_008771180.1 | 9 | 2442 | 139 | ||
XM_008772959.1 | XP_008771181.1 | 8 | 2251 | 139 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
