Assay Details
Gene Symbol: Arid5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244806.2 | XP_006244868.1 | 4 | 362 | 173 | |
XM_006244807.2 | XP_006244869.1 | 4 | 359 | 173 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
