Assay Details
Gene Symbol: Gpkow | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109381.1 | NP_001102851.1 | 11 | 3910 | 137 | |
GenBank mRNA | BC166817.1 | - | 9 | 3428 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P932059 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P074755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586072 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 161193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133038 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074756 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074760 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P932059 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P932059 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074756 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074760 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P074760 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P932059 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P074756 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P932059 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P389438 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15072 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103575 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10937084 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937086 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937081 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937082 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937085 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937083 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937079 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937088 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937087 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937078 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937080 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10937077 | - | - |
Rat Genome 230 2.0 Array | 1396280_at | - | - |
Rat Genome 230 2.0 Array | 1383255_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396280_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383255_at | - | - |
Affymetrix HT Rat Focus Array | 1396280_at | - | - |
Affymetrix HT Rat Focus Array | 1396280_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00009638.rn.1 | TC0X00001504.rn.1 | RTA TC |
Rat Expression Set 230 | 1396280_PM_at | - | - |
Rat Expression Set 230 | 1383255_PM_at | - | - |
Rat Expression Set 230 | 1383255_at | - | - |
Rat Expression Set 230 | 1396280_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10937077 | - | - |
Rat Genome U34 Set | rc_AI029480_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22070505 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357200 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362852 | - | - |
