Assay Details
Gene Symbol: Cdh19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009448.1 | NP_001009448.1 | 3 | 555 | 137 | |
XM_006249653.2 | XP_006249715.1 | 4 | 1193 | 137 | ||
GenBank mRNA | AB121032.1 | - | 3 | 555 | 137 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
