Assay Details
Gene Symbol: Hus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109092.1 | NP_001102562.1 | 8 | 1439 | 164 | |
XM_006251468.1 | XP_006251530.1 | 8 | 1434 | 164 | ||
XM_006251469.2 | XP_006251531.1 | 8 | 1638 | 164 | ||
XM_006251470.2 | XP_006251532.1 | 7 | 1310 | 164 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
