Assay Details
Gene Symbol: Dusp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007006.1 | NP_001007007.1 | 598 | 111 | ||
NM_001162408.1 | NP_001155880.1 | 1069 | 111 | |||
XM_008770542.1 | XP_008768764.1 | 4 - 5 | 1230 | 111 | ||
XM_008770543.1 | XP_008768765.1 | 1428 | 111 | |||
XM_008770544.1 | XP_008768766.1 | 3 - 4 | 1026 | 111 | ||
GenBank mRNA | BC083646.1 | - | 598 | 111 | ||
BC166532.1 | - | 1069 | 111 |
Gene Symbol: LOC100909469 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751425.3 | XP_003751473.2 | 4 - 5 | 636 | 111 | |
XM_003751426.3 | XP_003751474.1 | 2 - 3 | 1428 | 111 | ||
XM_008770474.1 | XP_008768696.1 | 4 - 5 | 1230 | 111 | ||
XM_008770475.1 | XP_008768697.1 | 3 - 4 | 1026 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
