Assay Details
Gene Symbol: Ldhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008893.1 | NP_001008893.1 | 5 - 6 | 669 | 135 | |
XM_006255631.2 | XP_006255693.1 | 5 - 6 | 720 | 135 | ||
GenBank mRNA | AY321341.1 | - | 5 - 6 | 669 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
