Assay Details
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009275.1 | NP_001009275.1 | 3 - 4 | 346 | 143 | |
GenBank mRNA | BC088178.1 | - | 3 - 4 | 346 | 143 | |
FQ209521.1 | - | 2 - 3 | 186 | 143 | ||
FQ223513.1 | - | 2 - 3 | 186 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
