Assay Details
Gene Symbol: Mrpl38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009369.2 | NP_001009369.2 | 7 - 8 | 879 | 131 | |
XM_008768372.1 | XP_008766594.1 | 7 - 8 | 1667 | 131 | ||
GenBank mRNA | BC087096.1 | - | 7 - 8 | 876 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
