Assay Details
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009539.1 | NP_001009539.1 | 5 - 6 | 962 | 154 | |
XM_006250253.2 | XP_006250315.1 | 5 - 6 | 993 | 154 | ||
XM_006250254.2 | XP_006250316.1 | 5 - 6 | 1113 | 154 | ||
GenBank mRNA | BC061812.1 | - | 5 - 6 | 962 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
