Assay Details
Gene Symbol: B4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009539.1 | NP_001009539.1 | 7 - 8 | 1192 | 129 | |
XM_006250253.2 | XP_006250315.1 | 7 - 8 | 1223 | 129 | ||
XM_006250254.2 | XP_006250316.1 | 7 - 8 | 1343 | 129 | ||
GenBank mRNA | BC061812.1 | - | 7 - 8 | 1192 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
