Assay Details
Gene Symbol: Zgpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009656.1 | NP_001009656.1 | 2 - 3 | 832 | 131 | |
XM_006235749.1 | XP_006235811.1 | 3 - 4 | 828 | 131 | ||
XM_006235750.1 | XP_006235812.1 | 3 - 4 | 793 | 131 | ||
XM_006235751.2 | XP_006235813.1 | 3 - 4 | 814 | 131 | ||
XM_006235752.2 | XP_006235814.1 | 2 - 3 | 728 | 131 | ||
GenBank mRNA | BC087720.1 | - | 2 - 3 | 832 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
