Assay Details
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009669.1 | NP_001009669.1 | 3 - 4 | 423 | 175 | |
GenBank mRNA | BC088757.1 | - | 3 - 4 | 423 | 175 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
